missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzenesulfonyl chloride, 99%

CAS: 832-53-1 | C6ClF5O2S | 266.57 g/mol

$488.83 - $488.83

Chemical Identifiers

CAS 832-53-1
Molecular Formula C6ClF5O2S
Molecular Weight (g/mol) 266.57
MDL Number MFCD00007427
InChI Key UOJCTEGNHXRPKO-UHFFFAOYSA-N
Synonym pentafluorobenzenesulfonyl chloride, 2,3,4,5,6-pentafluorobenzene-1-sulfonyl chloride, pentafluorobenzenesulphonyl chloride, benzenesulfonyl chloride, pentafluoro, pentafluorophenylsulfonyl chloride, 2,3,4,5,6-pentafluorobenzenesulfonyl choride, 2,3,4,5,6-pentafluoro-benzenesulfonyl chloride, chloro 2,3,4,5,6-pentafluorophenyl sulfone, pubchem5683, pentafluorobenzenesulfonylchloride
PubChem CID 70026
IUPAC Name 2,3,4,5,6-pentafluorobenzenesulfonyl chloride
SMILES C1(=C(C(=C(C(=C1F)F)S(=O)(=O)Cl)F)F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC147110250
SDS View Documents
Thermo Scientific Chemicals
147110250
25 g Glass bottle
Each for $488.83
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

832-53-1
266.57
UOJCTEGNHXRPKO-UHFFFAOYSA-N
70026
C1(=C(C(=C(C(=C1F)F)S(=O)(=O)Cl)F)F)F
C6ClF5O2S
MFCD00007427
pentafluorobenzenesulfonyl chloride, 2,3,4,5,6-pentafluorobenzene-1-sulfonyl chloride, pentafluorobenzenesulphonyl chloride, benzenesulfonyl chloride, pentafluoro, pentafluorophenylsulfonyl chloride, 2,3,4,5,6-pentafluorobenzenesulfonyl choride, 2,3,4,5,6-pentafluoro-benzenesulfonyl chloride, chloro 2,3,4,5,6-pentafluorophenyl sulfone, pubchem5683, pentafluorobenzenesulfonylchloride
2,3,4,5,6-pentafluorobenzenesulfonyl chloride

Specifications

832-53-1
211°C
98.5% min. (GC)
C6ClF5O2S
C6F5SO2Cl
25 g
pentafluorobenzenesulfonyl chloride, 2,3,4,5,6-pentafluorobenzene-1-sulfonyl chloride, pentafluorobenzenesulphonyl chloride, benzenesulfonyl chloride, pentafluoro, pentafluorophenylsulfonyl chloride, 2,3,4,5,6-pentafluorobenzenesulfonyl choride, 2,3,4,5,6-pentafluoro-benzenesulfonyl chloride, chloro 2,3,4,5,6-pentafluorophenyl sulfone, pubchem5683, pentafluorobenzenesulfonylchloride
C1(=C(C(=C(C(=C1F)F)S(=O)(=O)Cl)F)F)F
266.57
266.57
Pentafluorobenzenesulfonyl chloride, 99%
1.7960g/mL
90°C
Glass bottle
1.4780 to 1.48
MFCD00007427
1.796
UOJCTEGNHXRPKO-UHFFFAOYSA-N
2,3,4,5,6-pentafluorobenzenesulfonyl chloride
70026
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 212-620-0

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only