missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorobenzaldehyde 96.0+%, TCI America™
$313.16 - $1,252.65
Chemical Identifiers
| CAS | 653-37-2 |
|---|---|
| Molecular Formula | C7HF5O |
| Molecular Weight (g/mol) | 196.076 |
| MDL Number | MFCD00003303 |
| InChI Key | QJXCFMJTJYCLFG-UHFFFAOYSA-N |
| Synonym | pentafluorobenzaldehyde, perfluorobenzaldehyde, benzaldehyde, pentafluoro, benzaldehyde, 2,3,4,5,6-pentafluoro, unii-s9yie3jz4k, pentafluor-benzaldehyd, s9yie3jz4k, pubchem4253, pentafluoro-benzaldehyde, acmc-209nqh |
| PubChem CID | 69558 |
| IUPAC Name | 2,3,4,5,6-pentafluorobenzaldehyde |
| SMILES | C(=O)C1=C(C(=C(C(=C1F)F)F)F)F |
Chemical Identifiers
| 653-37-2 | |
| 196.076 | |
| QJXCFMJTJYCLFG-UHFFFAOYSA-N | |
| 69558 | |
| C(=O)C1=C(C(=C(C(=C1F)F)F)F)F |
| C7HF5O | |
| MFCD00003303 | |
| pentafluorobenzaldehyde, perfluorobenzaldehyde, benzaldehyde, pentafluoro, benzaldehyde, 2,3,4,5,6-pentafluoro, unii-s9yie3jz4k, pentafluor-benzaldehyd, s9yie3jz4k, pubchem4253, pentafluoro-benzaldehyde, acmc-209nqh | |
| 2,3,4,5,6-pentafluorobenzaldehyde |
Specifications
| 653-37-2 | |
| 170°C | |
| MFCD00003303 | |
| 2811 | |
| QJXCFMJTJYCLFG-UHFFFAOYSA-N | |
| 2,3,4,5,6-pentafluorobenzaldehyde | |
| 69558 | |
| ≥96.0% (GC) | |
| Pentafluorobenzaldehyde |
| White-Yellow | |
| C7HF5O | |
| 5 g | |
| pentafluorobenzaldehyde, perfluorobenzaldehyde, benzaldehyde, pentafluoro, benzaldehyde, 2,3,4,5,6-pentafluoro, unii-s9yie3jz4k, pentafluor-benzaldehyd, s9yie3jz4k, pubchem4253, pentafluoro-benzaldehyde, acmc-209nqh | |
| C(=O)C1=C(C(=C(C(=C1F)F)F)F)F | |
| 196.076 | |
| 196.08 | |
| Crystalline Powder |
Safety and Handling
TSCA : No