missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzaldehyde 96.0+%, TCI America™

Catalog No. P07465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P07465G 5 g
P074625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P07465G Supplier TCI America Supplier No. P07465G

Chemical Identifiers

CAS 653-37-2
Molecular Formula C7HF5O
Molecular Weight (g/mol) 196.076
MDL Number MFCD00003303
InChI Key QJXCFMJTJYCLFG-UHFFFAOYSA-N
Synonym pentafluorobenzaldehyde, perfluorobenzaldehyde, benzaldehyde, pentafluoro, benzaldehyde, 2,3,4,5,6-pentafluoro, unii-s9yie3jz4k, pentafluor-benzaldehyd, s9yie3jz4k, pubchem4253, pentafluoro-benzaldehyde, acmc-209nqh
PubChem CID 69558
IUPAC Name 2,3,4,5,6-pentafluorobenzaldehyde
SMILES C(=O)C1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Color White-Yellow
Boiling Point 170°C
Quantity 5 g
UN Number 2811
Formula Weight 196.08
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentafluorobenzaldehyde