missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoroanisole 98.0+%, TCI America™

Catalog No. p-7134843
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09185G 5 g
P091825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P09185G Supplier TCI America Supplier No. P09185G
Only null left

Chemical Identifiers

CAS 389-40-2
Molecular Formula C7H3F5O
Molecular Weight (g/mol) 198.092
MDL Number MFCD00000289
InChI Key ZRQUIRABLIQJRI-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluoroanisole, pentafluoroanisole, benzene, pentafluoromethoxy, pentafluorophenyl methyl ether, pentafluoromethoxybenzene, methyl pentafluorophenyl ether, benzene, 1,2,3,4,5-pentafluoro-6-methoxy, anisole, 2,3,4,5,6-pentafluoro, 1,2,3,4,6-pentafluoro-5-methoxybenzene, methoxypentafluorobenzene
PubChem CID 67849
IUPAC Name 1,2,3,4,5-pentafluoro-6-methoxybenzene
SMILES COC1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Color Colorless
Boiling Point 138°C
Quantity 5 g
UN Number 1993
Formula Weight 198.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentafluoroanisole