missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoro(phenoxy)cyclotriphosphazene 98.0+%, TCI America™

Catalog No. p-7135494
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22861G 1 g
P22865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22861G Supplier TCI America Supplier No. P22861G
Only null left

Chemical Identifiers

CAS 33027-68-8
Molecular Formula C6H5F5N3OP3
Molecular Weight (g/mol) 323.039
InChI Key XNZZEQCBAGUFMT-UHFFFAOYSA-N
Synonym 2,4,4,6,6-Pentafluoro-2-phenoxy-1,3,5,2,4,6-triazatriphosphorine
PubChem CID 23419251
IUPAC Name 2,2,4,4,6-pentafluoro-6-phenoxy-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
SMILES C1=CC=C(C=C1)OP2(=NP(=NP(=N2)(F)F)(F)F)F

Specifications

Color Colorless
Boiling Point 46°C
Quantity 1 g
UN Number 1760
Formula Weight 323.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentafluoro(phenoxy)cyclotriphosphazene
EINECSNumber (5)-6917
TSCA No
Recommended Storage Refrigerator