missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentaethylene Glycol 95.0+%, TCI America™
Supplier: TCI America P115425G
Specifications
| Pentaethylene Glycol | |
| Yellow | |
| C10H22O6 | |
| 25 g | |
| JLFNLZLINWHATN-UHFFFAOYSA-N | |
| 2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethanol | |
| 62551 | |
| 238.28 | |
| Liquid |
| 4792-15-8 | |
| 184°C | |
| MFCD00002878 | |
| pentaethylene glycol, 3,6,9,12-tetraoxatetradecane-1,14-diol, pentaethyleneglycol, pentaglycol, ho ch2ch2o 5h, 3,6,9,12-tetraoxatetradocane-1,14-diol, 2-2-2-2-2-hydroxyethoxy ethoxy ethoxy ethoxy ethanol, dsstox_cid_7579, dsstox_rid_78510, dsstox_gsid_27579 | |
| C(COCCOCCOCCOCCO)O | |
| 238.28 | |
| CHEBI:39631 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 4792-15-8 | |
| 238.28 | |
| JLFNLZLINWHATN-UHFFFAOYSA-N | |
| 62551 | |
| 2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethanol |
| C10H22O6 | |
| MFCD00002878 | |
| pentaethylene glycol, 3,6,9,12-tetraoxatetradecane-1,14-diol, pentaethyleneglycol, pentaglycol, ho ch2ch2o 5h, 3,6,9,12-tetraoxatetradocane-1,14-diol, 2-2-2-2-2-hydroxyethoxy ethoxy ethoxy ethoxy ethanol, dsstox_cid_7579, dsstox_rid_78510, dsstox_gsid_27579 | |
| CHEBI:39631 | |
| C(COCCOCCOCCOCCO)O |
Safety and Handling
EINECSNumber : (2)-0441
RTECSNumber : RZ2670000
TSCA : Yes