missing translation for 'onlineSavingsMsg'
Learn More

Pentaethylene Glycol 95.0+%, TCI America™

Catalog No. p-7134979
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11545G 5 g
P115425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11545G Supplier TCI America Supplier No. P11545G
Only null left

Chemical Identifiers

CAS 4792-15-8
Molecular Formula C10H22O6
Molecular Weight (g/mol) 238.28
MDL Number MFCD00002878
InChI Key JLFNLZLINWHATN-UHFFFAOYSA-N
Synonym pentaethylene glycol, 3,6,9,12-tetraoxatetradecane-1,14-diol, pentaethyleneglycol, pentaglycol, ho ch2ch2o 5h, 3,6,9,12-tetraoxatetradocane-1,14-diol, 2-2-2-2-2-hydroxyethoxy ethoxy ethoxy ethoxy ethanol, dsstox_cid_7579, dsstox_rid_78510, dsstox_gsid_27579
PubChem CID 62551
ChEBI CHEBI:39631
IUPAC Name 2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethanol
SMILES C(COCCOCCOCCOCCO)O

Specifications

Color Yellow
Boiling Point 184°C
Quantity 5 g
Formula Weight 238.28
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentaethylene Glycol
EINECSNumber (2)-0441
RTECSNumber RZ2670000
TSCA Yes