missing translation for 'onlineSavingsMsg'
Learn More

Pentaethylene Glycol Monomethyl Ether 96.0+%, TCI America™

Catalog No. p-7134982
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11591G 1 g
P11595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11591G Supplier TCI America Supplier No. P11591G
Only null left

Chemical Identifiers

CAS 23778-52-1
Molecular Formula C11H24O6
Molecular Weight (g/mol) 252.307
MDL Number MFCD00045995
InChI Key SLNYBUIEAMRFSZ-UHFFFAOYSA-N
Synonym mPEG5-Alcohol
PubChem CID 90255
IUPAC Name 2-[2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethoxy]ethanol
SMILES COCCOCCOCCOCCOCCO

Specifications

Color Yellow
Quantity 1 g
Formula Weight 252.31
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentaethylene Glycol Monomethyl Ether
EINECSNumber (7)-0097
TSCA Yes
Recommended Storage Refrigerator