accessibility menu, dialog, popup

UserName

H3COOOOOOH

CAS RN 23778-52-1

IUPAC Name: 2,5,8,11,14-pentaoxahexadecan-16-ol
Synonyms: 2,5,8,11,14-pentaoxahexadecan-16-ol
Molecular Weight (g/mol): 252.31
Molecular Formula: C11H24O6
InChi Key: SLNYBUIEAMRFSZ-UHFFFAOYSA-N
SMILES: COCCOCCOCCOCCOCCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results