missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentaethylene Glycol Monomethyl Ether 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 252.31 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Pentaethylene Glycol Monomethyl Ether |
Safety and Handling
| EINECSNumber | (7)-0097 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |