missing translation for 'onlineSavingsMsg'
Learn More

Pentaerythritol triacrylate, stab. with 4-methoxyphenol

Catalog No. AA1570536
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1570536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1570536 Supplier Thermo Scientific Chemicals Supplier No. 01570536

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3524-68-3 | C14H18O7 | 298.291 g/mol

Pentaerythritol triacrylate is used in UV-cured inks, dental polymers and varnishes. Further, it is used in the preparation of printing inks, coatings, in photoresists, acrylic glues, adhesives, binders, paper and wood impregnates. In addition, it serves as a modifier for polyester and fiberglass and plastic components for the medical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3524-68-3
Molecular Formula C14H18O7
Molecular Weight (g/mol) 298.291
MDL Number MFCD00009607
InChI Key HVVWZTWDBSEWIH-UHFFFAOYSA-N
Synonym pentaerythritol triacrylate, peta, pentaerythrityl triacrylate, gafgard 233, kayarad pet 30, sartomer sr 444, aronix m 305, setalux uv 2242, light acrylate pe 3a, p 300 acrylate
PubChem CID 19042
IUPAC Name [2-(hydroxymethyl)-3-prop-2-enoyloxy-2-(prop-2-enoyloxymethyl)propyl] prop-2-enoate
SMILES C=CC(=O)OCC(CO)(COC(=O)C=C)COC(=O)C=C

Specifications

Color Colorless
Density 1.16 g/mL
Flash Point >110°C (230°F)
Refractive Index 1.484
Quantity 500 g
Beilstein 6598752
Sensitivity Hygroscopic
Solubility Information Immiscible with water.
Formula Weight 298.28
Physical Form Liquid
Chemical Name or Material Pentaerythritol triacrylate, Stabilized with 300 to 400ppm 4-methoxyphenol
Show More Show Less
Hazard Category H315-H317-H319
Hazard Statement GHS H Statement
H302-H315-H319-H317
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c
EINECSNumber 222-540-8
RTECSNumber UD3370000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only