missing translation for 'onlineSavingsMsg'
Learn More

Pentaerythritol Tetrakis(mercaptoacetate) 95.0+%, TCI America™

Catalog No. p-7134830
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P088825G 25 g
P0888500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P088825G Supplier TCI America Supplier No. P088825G
Only null left

Chemical Identifiers

CAS 10193-99-4
Molecular Formula C13H20O8S4
Molecular Weight (g/mol) 432.535
MDL Number MFCD00022080
InChI Key RUDUCNPHDIMQCY-UHFFFAOYSA-N
Synonym Pentaerythritol Tetrathioglycolate
PubChem CID 82441
IUPAC Name [3-(2-sulfanylacetyl)oxy-2,2-bis[(2-sulfanylacetyl)oxymethyl]propyl] 2-sulfanylacetate
SMILES C(C(=O)OCC(COC(=O)CS)(COC(=O)CS)COC(=O)CS)S

Specifications

Color Colorless
Boiling Point 250°C
Quantity 25 g
Formula Weight 432.54
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Pentaerythritol Tetrakis(mercaptoacetate)
EINECSNumber (2)-3334
TSCA Yes