accessibility menu, dialog, popup

UserName

CAS RN 10193-99-4

IUPAC Name: 3-[(2-sulfanylacetyl)oxy]-2,2-bis({[(2-sulfanylacetyl)oxy]methyl})propyl 2-sulfanylacetate
Synonyms: 3-[(2-sulfanylacetyl)oxy]-2,2-bis({[(2-sulfanylacetyl)oxy]methyl})propyl 2-sulfanylacetate
Molecular Weight (g/mol): 432.54
Molecular Formula: C13H20O8S4
InChi Key: RUDUCNPHDIMQCY-UHFFFAOYSA-N
SMILES: SCC(=O)OCC(COC(=O)CS)(COC(=O)CS)COC(=O)CS
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results