missing translation for 'onlineSavingsMsg'
Learn More

Pentadecylbenzene 98.0+%, TCI America™

Catalog No. P066525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P066525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P066525ML Supplier TCI America Supplier No. P066525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2131-18-2
Molecular Formula C21H36
Molecular Weight (g/mol) 288.519
MDL Number MFCD00015083
InChI Key JIRNEODMTPGRGV-UHFFFAOYSA-N
Synonym 1-Phenylpentadecane
PubChem CID 16476
IUPAC Name pentadecylbenzene
SMILES CCCCCCCCCCCCCCCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 373°C
Quantity 25 mL
Formula Weight 288.52
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentadecylbenzene
EINECSNumber (3)-0021
TSCA Yes