accessibility menu, dialog, popup

UserName

H3C

CAS RN 2131-18-2

IUPAC Name: pentadecylbenzene
Synonyms: 1-phenylpentadecane
Molecular Weight (g/mol): 288.52
Molecular Formula: C21H36
InChi Key: JIRNEODMTPGRGV-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results