missing translation for 'onlineSavingsMsg'
Learn More

Pentadecane 99.0+%, TCI America™

Supplier:  TCI America P0606500ML

 View more versions of this product

Catalog No. P0606500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Pentadecane
Colorless
C15H32
500 mL
YCOZIPAWZNQLMR-UHFFFAOYSA-N
pentadecane
12391
212.42
Liquid
629-62-9
270°C
MFCD00008990
n-pentadecane, pentadekan, unii-16h6k2s8m2, ch3-ch2 13-ch3, mys, pentadecane, analytical standard, pentadecan, medicinal plant, pentadecyl group, pentadecane, n
CCCCCCCCCCCCCCC
212.42
CHEBI:28897
≥99.0% (GC)

Chemical Identifiers

629-62-9
212.42
YCOZIPAWZNQLMR-UHFFFAOYSA-N
12391
pentadecane
C15H32
MFCD00008990
n-pentadecane, pentadekan, unii-16h6k2s8m2, ch3-ch2 13-ch3, mys, pentadecane, analytical standard, pentadecan, medicinal plant, pentadecyl group, pentadecane, n
CHEBI:28897
CCCCCCCCCCCCCCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0010

RTECSNumber : RZ1800000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions