missing translation for 'onlineSavingsMsg'
Learn More

Pentadecane 99.0+%, TCI America™

Catalog No. p-7134694
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P060625ML 25 mL
P0606100ML 100 mL
P0606500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P060625ML Supplier TCI America Supplier No. P060625ML
Only null left

Chemical Identifiers

CAS 629-62-9
Molecular Formula C15H32
Molecular Weight (g/mol) 212.42
MDL Number MFCD00008990
InChI Key YCOZIPAWZNQLMR-UHFFFAOYSA-N
Synonym n-pentadecane, pentadekan, unii-16h6k2s8m2, ch3-ch2 13-ch3, mys, pentadecane, analytical standard, pentadecan, medicinal plant, pentadecyl group, pentadecane, n
PubChem CID 12391
ChEBI CHEBI:28897
IUPAC Name pentadecane
SMILES CCCCCCCCCCCCCCC

Specifications

Color Colorless
Boiling Point 270°C
Quantity 25 mL
Formula Weight 212.42
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentadecane
EINECSNumber (2)-0010
RTECSNumber RZ1800000
TSCA Yes