missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentadecafluorotriethylamine 95.0+%, TCI America™
Supplier: TCI America P134825G
Specifications
| Pentadecafluorotriethylamine | |
| Yellow | |
| C6F15N | |
| 25 g | |
| Perfluorotriethylamine, Tris(pentafluoroethyl)amine | |
| FC(F)(F)C(F)(F)N(C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)F | |
| 371.05 | |
| 371.05 | |
| Liquid |
| 359-70-6 | |
| 72°C | |
| MFCD00166270 | |
| 3267 | |
| CBEFDCMSEZEGCX-UHFFFAOYSA-N | |
| tris(1,1,2,2,2-pentafluoroethyl)amine | |
| 78986 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 359-70-6 | |
| 371.05 | |
| CBEFDCMSEZEGCX-UHFFFAOYSA-N | |
| 78986 | |
| FC(F)(F)C(F)(F)N(C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)F |
| C6F15N | |
| MFCD00166270 | |
| Perfluorotriethylamine, Tris(pentafluoroethyl)amine | |
| tris(1,1,2,2,2-pentafluoroethyl)amine |
Safety and Handling
RTECSNumber : KH2155170
TSCA : No