accessibility menu, dialog, popup

UserName

CAS RN 359-70-6

IUPAC Name: tris(1,1,2,2,2-pentafluoroethyl)amine
Synonyms: perfluoro-tri-n-ethylamine
Molecular Weight (g/mol): 371.05
Molecular Formula: C6F15N
InChi Key: CBEFDCMSEZEGCX-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)N(C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results