missing translation for 'onlineSavingsMsg'
Learn More

Pentadecafluorotriethylamine 95.0+%, TCI America™

Catalog No. P13485G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13485G 5 g
P134825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13485G Supplier TCI America Supplier No. P13485G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 359-70-6
Molecular Formula C6F15N
Molecular Weight (g/mol) 371.05
MDL Number MFCD00166270
InChI Key CBEFDCMSEZEGCX-UHFFFAOYSA-N
Synonym Perfluorotriethylamine, Tris(pentafluoroethyl)amine
PubChem CID 78986
IUPAC Name tris(1,1,2,2,2-pentafluoroethyl)amine
SMILES FC(F)(F)C(F)(F)N(C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 72°C
Quantity 5 g
UN Number 3267
Formula Weight 371.05
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentadecafluorotriethylamine
RTECSNumber KH2155170
TSCA No