missing translation for 'onlineSavingsMsg'
Learn More

Pentacontane 98.0+%, TCI America™

Catalog No. P09641G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09641G 1 g
P0964100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P09641G Supplier TCI America Supplier No. P09641G
Only null left

Chemical Identifiers

CAS 6596-40-3
Molecular Formula C50H102
Molecular Weight (g/mol) 703.366
MDL Number MFCD00042907
InChI Key PFLUOWJPZLHUEA-UHFFFAOYSA-N
PubChem CID 81067
IUPAC Name pentacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Physical Form Powder/Crystals
Color White
Melting Point 94°C
Quantity 1 g
Formula Weight 703.37
Percent Purity ≥98.0% (GC)
Chemical Name or Material Pentacontane