missing translation for 'onlineSavingsMsg'
Learn More

Pentachloropyridine 98.0+%, TCI America™

Catalog No. P166125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P166125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P166125G Supplier TCI America Supplier No. P166125G
Only null left

Chemical Identifiers

CAS 2176-62-7
Molecular Formula C5Cl5N
Molecular Weight (g/mol) 251.312
InChI Key DNDPLEAVNVOOQZ-UHFFFAOYSA-N
Synonym pentachloropyridine, perchloropyridine, pyridine, pentachloro, pyridine, 2,3,4,5,6-pentachloro, unii-6mm852ka6t, 2,3,4,5,6-pentachlorpyridine, dsstox_cid_2179, dsstox_rid_76515, dsstox_gsid_22179, pentachlorpyridine
PubChem CID 16584
IUPAC Name 2,3,4,5,6-pentachloropyridine
SMILES C1(=C(C(=NC(=C1Cl)Cl)Cl)Cl)Cl

Specifications

Melting Point 126°C
Color White
Boiling Point 280°C
Quantity 25 g
Formula Weight 251.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentachloropyridine
RTECSNumber UT7000000
TSCA Yes