CAS RN 2176-62-7
CAS RN 2176-62-7
IUPAC Name:
pentachloropyridine
Synonyms:
2,3,4,5,6-pentachlorpyridine
Molecular Weight (g/mol):
251.31
Molecular Formula:
C5Cl5N
InChi Key:
DNDPLEAVNVOOQZ-UHFFFAOYSA-N
SMILES:
ClC1=NC(Cl)=C(Cl)C(Cl)=C1Cl
1
–
1
of
1
results
1
Pentachloropyridine 98.0+%, TCI America™
CAS: 2176-62-7 Molecular Formula: C5Cl5N Molecular Weight (g/mol): 251.312 InChI Key: DNDPLEAVNVOOQZ-UHFFFAOYSA-N Synonym: pentachloropyridine,perchloropyridine,pyridine, pentachloro,pyridine, 2,3,4,5,6-pentachloro,unii-6mm852ka6t,2,3,4,5,6-pentachlorpyridine,dsstox_cid_2179,dsstox_rid_76515,dsstox_gsid_22179,pentachlorpyridine PubChem CID: 16584 IUPAC Name: 2,3,4,5,6-pentachloropyridine SMILES: C1(=C(C(=NC(=C1Cl)Cl)Cl)Cl)Cl