missing translation for 'onlineSavingsMsg'
Learn More

Pentachlorophenol, MilliporeSigma™ Supelco™

Catalog No. 111016697
Change view
Click to view available options
Quantity:
5000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016697 5000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016697 Supplier Merck Emd Millipore Supplier No. 48555U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pentachlorophenol (PCP) is a chlorinated hydrocarbon, an important industrial chemical and a general biocide. It is primarily used in wood protection.

Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Packaging Ampule
CAS 87-86-5
Vapor Pressure 40 mmHg (211.2°C)
Vapor Density 9.2 (vs. air)
Density 1.978 g/mL (at 25°C)
Physical Form Neat
Linear Formula C6 Cl5 OH
MDL Number MFCD00002162
Melting Point 165°C to 180°C
Boiling Point 310°C
Type Food and Cosmetic Component Standard
Quantity 5000 mg
Formula Weight 266.34
Grade Analytical Standard
Show More Show Less