missing translation for 'onlineSavingsMsg'
Learn More

Pentacene (purified by sublimation), TCI America™

Catalog No. P0030100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0030100MG 100 mg
P00301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0030100MG Supplier TCI America Supplier No. P0030100MG

Chemical Identifiers

CAS 135-48-8
Molecular Formula C22H14
Molecular Weight (g/mol) 278.35
MDL Number MFCD00003710
InChI Key SLIUAWYAILUBJU-UHFFFAOYSA-N
Synonym benzo b naphthacene, lin-dibenzanthracene, lin-naphthoanthracene, 2,3:6,7-dibenzanthracene, unii-9fqu5ha0uy, 9fqu5ha0uy, pentacen-2-yl, pentacene, 2,7-dibenzanthracene
PubChem CID 8671
ChEBI CHEBI:33148
IUPAC Name pentacene
SMILES C1=CC2=CC3=CC4=CC5=CC=CC=C5C=C4C=C3C=C2C=C1

Specifications

Color Black-Purple
Quantity 100 mg
Formula Weight 278.35
Physical Form Crystalline Powder
Chemical Name or Material Pentacene (purified by sublimation)
RTECSNumber RY9400000
TSCA Yes