missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentabromobenzyl Acrylate 98.0+%, TCI America™
$158.44 - $521.94
Chemical Identifiers
| CAS | 59447-55-1 |
|---|---|
| Molecular Formula | C10H5Br5O2 |
| Molecular Weight (g/mol) | 556.668 |
| MDL Number | MFCD00804467 |
| InChI Key | GRKDVZMVHOLESV-UHFFFAOYSA-N |
| Synonym | Acrylic Acid Pentabromobenzyl Ester |
| PubChem CID | 101059 |
| IUPAC Name | (2,3,4,5,6-pentabromophenyl)methyl prop-2-enoate |
| SMILES | C=CC(=O)OCC1=C(C(=C(C(=C1Br)Br)Br)Br)Br |
Chemical Identifiers
| 59447-55-1 | |
| 556.668 | |
| GRKDVZMVHOLESV-UHFFFAOYSA-N | |
| 101059 | |
| C=CC(=O)OCC1=C(C(=C(C(=C1Br)Br)Br)Br)Br |
| C10H5Br5O2 | |
| MFCD00804467 | |
| Acrylic Acid Pentabromobenzyl Ester | |
| (2,3,4,5,6-pentabromophenyl)methyl prop-2-enoate |
Specifications
| 59447-55-1 | |
| Yellow | |
| MFCD00804467 | |
| Acrylic Acid Pentabromobenzyl Ester | |
| C=CC(=O)OCC1=C(C(=C(C(=C1Br)Br)Br)Br)Br | |
| 556.668 | |
| 556.67 | |
| Crystalline Powder |
| 125°C | |
| C10H5Br5O2 | |
| 1 g | |
| GRKDVZMVHOLESV-UHFFFAOYSA-N | |
| (2,3,4,5,6-pentabromophenyl)methyl prop-2-enoate | |
| 101059 | |
| ≥98.0% (GC) | |
| Pentabromobenzyl Acrylate |
Safety and Handling
EINECSNumber : (3)-4183
TSCA : Yes
Recommended Storage : Refrigerator