accessibility menu, dialog, popup

UserName

BrBrBrOOCH2BrBr

CAS RN 59447-55-1

IUPAC Name: (2,3,4,5,6-pentabromophenyl)methyl prop-2-enoate
Synonyms: (2,3,4,5,6-pentabromophenyl)methyl prop-2-enoate
Molecular Weight (g/mol): 556.67
Molecular Formula: C10H5Br5O2
InChi Key: GRKDVZMVHOLESV-UHFFFAOYSA-N
SMILES: BrC1=C(Br)C(Br)=C(COC(=O)C=C)C(Br)=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results