missing translation for 'onlineSavingsMsg'
Learn More

Pentabromobenzyl Acrylate 98.0+%, TCI America™

Catalog No. P18775G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18775G 5 g
P18771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18775G Supplier TCI America Supplier No. P18775G

Chemical Identifiers

CAS 59447-55-1
Molecular Formula C10H5Br5O2
Molecular Weight (g/mol) 556.668
MDL Number MFCD00804467
InChI Key GRKDVZMVHOLESV-UHFFFAOYSA-N
Synonym Acrylic Acid Pentabromobenzyl Ester
PubChem CID 101059
IUPAC Name (2,3,4,5,6-pentabromophenyl)methyl prop-2-enoate
SMILES C=CC(=O)OCC1=C(C(=C(C(=C1Br)Br)Br)Br)Br

Specifications

Melting Point 125°C
Color Yellow
Quantity 5 g
Formula Weight 556.67
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentabromobenzyl Acrylate
EINECSNumber (3)-4183
TSCA Yes
Recommended Storage Refrigerator