missing translation for 'onlineSavingsMsg'
Learn More

Penicillin V potassium salt

Catalog No. AAJ6244209
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6244209 10 g
AAJ6244214 25 g
AAJ6244222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6244209 Supplier Thermo Scientific Chemicals Supplier No. J6244209

Inhibits bacterial cell wall synthesis | CAS: 132-98-9 | C16H17KN2O5S | 388.48 g/mol

Penicillin V Potassium bactericidal against penicillin-susceptible microorganisms during the stage of active multiplication. It inhibits biosynthesis of cell-wall mucopeptide. It is used to treat or prevent infections that are proven or strongly suspected to be caused by bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Penicillin V Potassium bactericidal against penicillin-susceptible microorganisms during the stage of active multiplication. It inhibits biosynthesis of cell-wall mucopeptide. It is used to treat or prevent infections that are proven or strongly suspected to be caused by bacteria.

Solubility
Soluble in water, dimethyl sulfoxide, and methanol.

Notes
Store at -20°C. Incompatible with oxidizing agents. Protect from heat.

Chemical Identifiers

CAS 132-98-9
Molecular Formula C16H17KN2O5S
Molecular Weight (g/mol) 388.48
MDL Number MFCD00051771
InChI Key HCTVWSOKIJULET-LQDWTQKMSA-M
Synonym Phenoxymethylpenicillinic acid potassium salt
PubChem CID 131673927
IUPAC Name (2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenoxyacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;potassium
SMILES [K+].CC1(C)S[C@@H]2[C@H](NC(=O)COC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O

Specifications

Type Penicillin V Potassium Salt
Physical Form Powder
Melting Point 197°C to 202°C
Color White
Quantity 10 g
Beilstein 3899451
Merck Index 14,7100
Solubility Information Soluble in water,dimethyl sulfoxide,and methanol.
Formula Weight 388.48
Chemical Name or Material Penicillin V potassium salt
Hazard Category H302-H317-H334
Hazard Statement GHS H Statement
H334-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280g-P285-P301+P312-P302+P352-P304+P341-P330-P333+P313-P342+P311-P363-P501c
EINECSNumber 205-086-5
RTECSNumber XH9275000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only