missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Penicillin G Sodium Salt 98.0+%, TCI America™
$72.15 - $246.82
Chemical Identifiers
| CAS | 69-57-8 |
|---|---|
| Molecular Formula | C16H17N2NaO4S |
| Molecular Weight (g/mol) | 356.372 |
| MDL Number | MFCD00069666 |
| InChI Key | FCPVYOBCFFNJFS-LQDWTQKMSA-M |
| Synonym | Benzylpenicillin Sodium Salt |
| PubChem CID | 23668834 |
| ChEBI | CHEBI:51765 |
| IUPAC Name | sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate |
| SMILES | CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+] |
Chemical Identifiers
| 69-57-8 | |
| 356.372 | |
| FCPVYOBCFFNJFS-LQDWTQKMSA-M | |
| 23668834 | |
| sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate |
| C16H17N2NaO4S | |
| MFCD00069666 | |
| Benzylpenicillin Sodium Salt | |
| CHEBI:51765 | |
| CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+] |
Specifications
| Penicillin G Sodium Salt | |
| 69-57-8 | |
| White | |
| MFCD00069666 | |
| FCPVYOBCFFNJFS-LQDWTQKMSA-M | |
| sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate | |
| 23668834 | |
| 356.37 | |
| Penicillin G Sodium Salt |
| Crystalline Powder | |
| 227°C | |
| C16H17N2NaO4S | |
| Benzylpenicillin Sodium Salt | |
| CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+] | |
| 356.372 | |
| CHEBI:51765 | |
| ≥98.0% (HPLC,N) |
Safety and Handling
RTECSNumber : XH9800000
TSCA : No