missing translation for 'onlineSavingsMsg'
Learn More

Penicillin G Sodium Salt 98.0+%, TCI America™

Catalog No. P17705G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17705G 5 g
P17701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17705G Supplier TCI America Supplier No. P17705G

Chemical Identifiers

CAS 69-57-8
Molecular Formula C16H17N2NaO4S
Molecular Weight (g/mol) 356.372
MDL Number MFCD00069666
InChI Key FCPVYOBCFFNJFS-LQDWTQKMSA-M
Synonym Benzylpenicillin Sodium Salt
PubChem CID 23668834
ChEBI CHEBI:51765
IUPAC Name sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+]

Specifications

Type Penicillin G Sodium Salt
Physical Form Crystalline Powder
Melting Point 227°C
Color White
Quantity 5 g
Formula Weight 356.37
Percent Purity ≥98.0% (HPLC,N)
Chemical Name or Material Penicillin G Sodium Salt
RTECSNumber XH9800000
TSCA No