missing translation for 'onlineSavingsMsg'
Learn More

Penicillin G potassium salt

Catalog No. p-7042828
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6390114 25 g
AAJ6390122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6390114 Supplier Thermo Scientific Chemicals Supplier No. J6390114
Only null left

Inhibits bacterial cell wall synthesis | CAS: 113-98-4 | C33H36ClKN4O4S2 | 691.34 g/mol

Penicillin G potassium salt is used as a cell culture additive as an antibiotics. Use to inhibit the synthesis of bacterial cell walls by inhibition of the cell wall peptidoglycan chain cross-lining. It is the drug of choice for groups A, B, C and G streptococci, nonenterococcal group D streptococci, viridians group streptococci, and non-penicillinase producing staphylococcus. The potassium salt has been used to study murosomes of staphylococci and the penicillin-induced lysis of Streptococcus mutans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Penicillin G potassium salt is used as a cell culture additive as an antibiotics. Use to inhibit the synthesis of bacterial cell walls by inhibition of the cell wall peptidoglycan chain cross-lining. It is the drug of choice for groups A, B, C and G streptococci, nonenterococcal group D streptococci, viridians group streptococci, and non-penicillinase producing staphylococcus. The potassium salt has been used to study murosomes of staphylococci and the penicillin-induced lysis of Streptococcus mutans.

Solubility
Soluble in water (100 mg/ml), methanol, ethanol (sparingly), and alcohol. Insoluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 113-98-4
Molecular Formula C33H36ClKN4O4S2
Molecular Weight (g/mol) 691.34
MDL Number MFCD00036193
InChI Key HTQQVOYPSGNVPA-HBRCYENSSA-M
Synonym Benzylpenicillin potassium salt; Potassium benzylpenicillinate
PubChem CID 23664709
ChEBI CHEBI:7963
IUPAC Name potassium [3-(2-chloro-10H-phenothiazin-10-yl)propyl]dimethylamine (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [K+].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(Cl)C=C12.CC1(C)S[C@@H]2[C@H](NC(=O)CC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O

Specifications

Type Penicillin G Potassium Salt
Physical Form Powder
Color White
Quantity 25 g
Beilstein 3832841
Merck Index 14,7094
Solubility Information Soluble in water (100mg/ml),methanol,ethanol (sparingly),and alcohol. Insoluble in chloroform.
Formula Weight 372.48
Chemical Name or Material Penicillin G potassium salt
Hazard Category H317-H334
Hazard Statement GHS H Statement
H334-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
EINECSNumber 204-038-0
RTECSNumber XH9700000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only