missing translation for 'onlineSavingsMsg'
Learn More

Penfluridol 98.0+%, TCI America™

Supplier:  TCI America P20761G

 View more versions of this product

Catalog No. P20761G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Penfluridol
108°C
C28H27ClF5NO
1 g
1-[4,4-Bis(4-fluorophenyl)butyl]-4-[4-chloro-3-(trifluoromethyl)phenyl]-4-piperidinol
C1CN(CCC1(C2=CC(=C(C=C2)Cl)C(F)(F)F)O)CCCC(C3=CC=C(C=C3)F)C4=CC=C(C=C4)F
523.972
523.97
Crystalline Powder
26864-56-2
White-Yellow
MFCD00866714
2811
MDLAAYDRRZXJIF-UHFFFAOYSA-N
1-[4,4-bis(4-fluorophenyl)butyl]-4-[4-chloro-3-(trifluoromethyl)phenyl]piperidin-4-ol
33630
≥98.0% (T)

Chemical Identifiers

26864-56-2
523.972
MDLAAYDRRZXJIF-UHFFFAOYSA-N
33630
C1CN(CCC1(C2=CC(=C(C=C2)Cl)C(F)(F)F)O)CCCC(C3=CC=C(C=C3)F)C4=CC=C(C=C4)F
C28H27ClF5NO
MFCD00866714
1-[4,4-Bis(4-fluorophenyl)butyl]-4-[4-chloro-3-(trifluoromethyl)phenyl]-4-piperidinol
1-[4,4-bis(4-fluorophenyl)butyl]-4-[4-chloro-3-(trifluoromethyl)phenyl]piperidin-4-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : TN6957000

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions