missing translation for 'onlineSavingsMsg'
Learn More

Penfluridol 98.0+%, TCI America™

Catalog No. P2076200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2076200MG 200 mg
P20761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2076200MG Supplier TCI America Supplier No. P2076200MG
Only null left

Chemical Identifiers

CAS 26864-56-2
Molecular Formula C28H27ClF5NO
Molecular Weight (g/mol) 523.972
MDL Number MFCD00866714
InChI Key MDLAAYDRRZXJIF-UHFFFAOYSA-N
Synonym 1-[4,4-Bis(4-fluorophenyl)butyl]-4-[4-chloro-3-(trifluoromethyl)phenyl]-4-piperidinol
PubChem CID 33630
IUPAC Name 1-[4,4-bis(4-fluorophenyl)butyl]-4-[4-chloro-3-(trifluoromethyl)phenyl]piperidin-4-ol
SMILES C1CN(CCC1(C2=CC(=C(C=C2)Cl)C(F)(F)F)O)CCCC(C3=CC=C(C=C3)F)C4=CC=C(C=C4)F

Specifications

Melting Point 108°C
Color White-Yellow
Quantity 200 mg
UN Number 2811
Formula Weight 523.97
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Penfluridol
RTECSNumber TN6957000
TSCA No
Recommended Storage Refrigerator