missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pendimethalin 98.0+%, TCI America™
$188.67 - $646.85
Chemical Identifiers
| CAS | 40487-42-1 |
|---|---|
| Molecular Formula | C13H19N3O4 |
| Molecular Weight (g/mol) | 281.312 |
| MDL Number | MFCD00055332 |
| InChI Key | CHIFOSRWCNZCFN-UHFFFAOYSA-N |
| Synonym | pendimethalin, pendimethaline, penoxaline, prowl, herbadox, penoxalin, phenoxalin, accotab, stomp, way up |
| PubChem CID | 38479 |
| ChEBI | CHEBI:83569 |
| IUPAC Name | 3,4-dimethyl-2,6-dinitro-N-pentan-3-ylaniline |
| SMILES | CCC(CC)NC1=C(C=C(C(=C1[N+](=O)[O-])C)C)[N+](=O)[O-] |
Chemical Identifiers
| 40487-42-1 | |
| 281.312 | |
| CHIFOSRWCNZCFN-UHFFFAOYSA-N | |
| 38479 | |
| 3,4-dimethyl-2,6-dinitro-N-pentan-3-ylaniline |
| C13H19N3O4 | |
| MFCD00055332 | |
| pendimethalin, pendimethaline, penoxaline, prowl, herbadox, penoxalin, phenoxalin, accotab, stomp, way up | |
| CHEBI:83569 | |
| CCC(CC)NC1=C(C=C(C(=C1[N+](=O)[O-])C)C)[N+](=O)[O-] |
Specifications
| 40487-42-1 | |
| C13H19N3O4 | |
| 5 g | |
| pendimethalin, pendimethaline, penoxaline, prowl, herbadox, penoxalin, phenoxalin, accotab, stomp, way up | |
| CCC(CC)NC1=C(C=C(C(=C1[N+](=O)[O-])C)C)[N+](=O)[O-] | |
| 281.312 | |
| CHEBI:83569 | |
| ≥98.0% (GC) | |
| Pendimethalin |
| 58°C | |
| MFCD00055332 | |
| 3077 | |
| CHIFOSRWCNZCFN-UHFFFAOYSA-N | |
| 3,4-dimethyl-2,6-dinitro-N-pentan-3-ylaniline | |
| 38479 | |
| 281.31 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : BX5470000
TSCA : No