missing translation for 'onlineSavingsMsg'
Learn More

Penciclovir 98.0+%, TCI America™

Supplier:  TCI America P21641G

 View more versions of this product

Catalog No. P21641G


Only null left

Description

Description

Specifications

Specifications

Penciclovir
Powder/Crystals
MFCD15142866
278°C
JNTOCHDNEULJHD-UHFFFAOYSA-N
2-amino-9-[4-hydroxy-3-(hydroxymethyl)butyl]-3H-purin-6-one
1 g
CHEBI:7956
≥98.0% (HPLC,T)
39809-25-1
C10H15N5O3
White
BRL 39123, 2-Amino-9-[4-hydroxy-3-(hydroxymethyl)butyl]-1H-purin-6-one, 9-[4-Hydroxy-3-(hydroxymethyl)butyl]guanine
C1=NC2=C(N1CCC(CO)CO)NC(=NC2=O)N
253.262
4725
253.26

Chemical Identifiers

39809-25-1
253.262
JNTOCHDNEULJHD-UHFFFAOYSA-N
4725
2-amino-9-[4-hydroxy-3-(hydroxymethyl)butyl]-3H-purin-6-one
C10H15N5O3
MFCD15142866
BRL 39123, 2-Amino-9-[4-hydroxy-3-(hydroxymethyl)butyl]-1H-purin-6-one, 9-[4-Hydroxy-3-(hydroxymethyl)butyl]guanine
CHEBI:7956
C1=NC2=C(N1CCC(CO)CO)NC(=NC2=O)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : UP0789400

TSCA : No

Recommended Storage : Freezer (-20 °C)

SDS
Documents

Documents

Promotions

Promotions