missing translation for 'onlineSavingsMsg'
Learn More

Penciclovir 98.0+%, TCI America™

Catalog No. P2164200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2164200MG 200 mg
P21641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2164200MG Supplier TCI America Supplier No. P2164200MG

Chemical Identifiers

CAS 39809-25-1
Molecular Formula C10H15N5O3
Molecular Weight (g/mol) 253.262
MDL Number MFCD15142866
InChI Key JNTOCHDNEULJHD-UHFFFAOYSA-N
Synonym BRL 39123, 2-Amino-9-[4-hydroxy-3-(hydroxymethyl)butyl]-1H-purin-6-one, 9-[4-Hydroxy-3-(hydroxymethyl)butyl]guanine
PubChem CID 4725
ChEBI CHEBI:7956
IUPAC Name 2-amino-9-[4-hydroxy-3-(hydroxymethyl)butyl]-3H-purin-6-one
SMILES C1=NC2=C(N1CCC(CO)CO)NC(=NC2=O)N

Specifications

Physical Form Powder/Crystals
Color White
Melting Point 278°C
Quantity 200 mg
Formula Weight 253.26
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Penciclovir
RTECSNumber UP0789400
TSCA No
Recommended Storage Freezer (-20 °C)