missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PECR Polyclonal Antibody

Description
Immunogen sequence: EEEVNNLVKS TLDTFGKINF LVNNGGGQFL SPAEHISSKG WHAVLETNLT GTFYMCKAVY SSWMKEHGGS IVNIIVPTKA GFPLAVHSGA ARAGVYNLTK SLA Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 80%.
PECR (Peroxisomal Trans-2-Enoyl-CoA Reductase) is a Protein Coding gene. Among its related pathways are Metabolism and Allograft rejection. Gene Ontology (GO) annotations related to this gene include signaling receptor binding and coenzyme binding. An important paralog of this gene is DECR1.
Specifications
Specifications
| Antigen | PECR |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Pecr |
| Gene Accession No. | Q9BY49 |
| Gene Alias | 2,4-dienoyl-CoA reductase-related protein; 2400003B18Rik; DCRRP; DCR-RP; HPDHase; HSA250303; PECR; perosisomal 2-enoyl-CoA reductase; peroxisomal 2,4-dienoyl-CoA reductase; Peroxisomal trans-2-enoyl-CoA reductase; PRO1004; putative short chain alcohol dehydrogenase; PVIARL; pVI-ARL; PX-2,4-DCR1; RLF98; SDR29C1; Short chain dehydrogenase/reductase family 29C member 1; short chain dehydrogenase/reductase family 29C, member 1; TERP |
| Gene Symbols | Pecr |
| Show More |