missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NoTrax™ Pebble Trax™

Pebble Trax™ is an anti-fatigue floor mat for assembly lines, lab areas, machine workstations and packing areas
$149.92 - $899.55
Specifications
| Color | Black, Yellow |
|---|---|
| Material | Top Surface Dense Closed Cell PVC Foam Base |
| Type | Anti-fatigue Floor Mat |
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19085912
![]() |
NoTrax™
480S0023BL |
2 x 3 ft. |
Each for $149.92
|
|
|||||
19085913
![]() |
NoTrax™
480S0035BL |
3 x 5 ft. |
Each for $374.80
|
|
|||||
19085914
![]() |
NoTrax™
480S0312BL |
3 x 12 ft. |
Each for $899.55
|
|
|||||
Description
- Anti-fatigue floor mat offers comfort and ergonomic support
- For dry environments like assembly lines, lab areas, machine workstations and packing areas
- Pebble-embossed rubber top provides non-directional traction
- Easy to clean
- Resists exposure to sparks from welding
- Made with a resilient SBR rubber compound, top surface combined with dense closed cell foam base utilizing Notrax™ exclusive UniFusion™ technology
- Eliminates the possibility of de-lamination
- Comes standard with RedStop™
- Uniquely engineered non-slip backing technology eliminates slipping and sliding of floor mats on smooth surface flooring such as tile, wood, marble, and treated concrete
- Sloped beveled edges reduce tripping hazards and allow easy access on and off mat
Specifications
| Black, Yellow | |
| Top Surface Dense Closed Cell PVC Foam Base | |
| Anti-fatigue Floor Mat |