missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDZ-GEF1 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories Inc A301966A
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 or Tris-EDTA pH 9.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Dog
Functions as a guanine nucleotide exchange factor (GEF), which activates Rap and Ras family of small GTPases by exchanging bound GDP for free GTP in a cAMP-dependent manner. Serves as a link between cell surface receptors and Rap/Ras GTPases in intracellular signaling cascades. Acts also as an effector for Rap1 by direct association with Rap1-GTP thereby leading to the amplification of Rap1-mediated signaling. Shows weak activity on HRAS. It is controversial whether RAPGEF2 binds cAMP and cGMP (PubMed:23800469, PubMed:10801446) or not (PubMed:10608844, PubMed:10548487, PubMed:11359771). Its binding to ligand-activated beta-1 adrenergic receptor ADRB1 leads to the Ras activation through the G(s)-alpha signaling pathway. Involved in the cAMP- induced Ras and Erk1/2 signaling pathway that leads to sustained inhibition of long term melanogenesis by reducing dendrite extension and melanin synthesis. Provides also inhibitory signals for cell proliferation of melanoma cells and promotes their apoptosis in a cAMP-independent ner. Regulates cAMP-induced neuritogenesis by mediating the Rap1/B-Raf/ERK signaling through a pathway that is independent on both PKA and RAPGEF3/RAPGEF4. Involved in neuron migration and in the formation of the major forebrain fiber connections forming the corpus callosum, the anterior commissure and the hippocampal commissure during brain development. Involved in neuronal growth factor (NGF)-induced sustained activation of Rap1 at late endosomes and in brain- derived neurotrophic factor (BDNF)-induced axon outgrowth of hippocampal neurons. Plays a role in the regulation of embryonic blood vessel formation and in the establishment of basal junction integrity and endothelial barrier function. May be involved in the regulation of the vascular endothelial growth factor receptor KDR and cadherin CDH5 expression at allantois endothelial cell-cell junctions.Chemical Identifiers
| PDZ-GEF1 | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| IgG | |
| 4° C |
| 1 mg/ml | |
| Polyclonal | |
| Liquid | |
| RUO | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Antigen affinity chromatography | |
| Antibody |
Specifications
| PDZ-GEF1 | |
| Polyclonal | |
| Unconjugated | |
| RAPGEF2 | |
| Rap guanine nucleotide exchange factor 2, Cyclic nucleotide ras GEF, CNrasGEF, Neural RAP guanine nucleotide exchange protein, nRap GEP, PDZ domain-containing guanine nucleotide exchange factor 1, PDZ-GEF1, RA-GEF-1, Ras/Rap1-associating GEF-1, RAPGEF2, KIAA0313, NRAPGEP, PDZGEF1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 76089, 9693 | |
| 4° C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q8CHG7, Q9Y4G8 | |
| RAPGEF2 | |
| Between 975 and 1025 | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |