missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDXDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112232
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112232 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112232 Supplier Invitrogen™ Supplier No. PA5112232
Only null left

Rabbit Polyclonal Antibody

PDXDC1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP

PDXDC1 (Pyridoxal Dependent Decarboxylase Domain Containing 1) is a Protein Coding gene. Diseases associated with PDXDC1 include Autosomal Recessive Nonsyndromic Deafness 32 and Fetal Akinesia Deformation Sequence 1. Gene Ontology (GO) annotations related to this gene include pyridoxal phosphate binding and carboxy-lyase activity. An important paralog of this gene is HDC. [GeneCards].

Specifications

Antigen PDXDC1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene Pdxdc1
Gene Accession No. Q6P996
Gene Alias 2210010A19Rik; AA415817; KIAA0251; Kiaa0251-hp; LP8165; Pdxdc1; pyridoxal dependent decarboxylase domain containing 1; pyridoxal-dependent decarboxylase domain containing 1; pyridoxal-dependent decarboxylase domain-containing protein 1; RGD1562597
Gene Symbols Pdxdc1
Host Species Rabbit
Immunogen E. coli-derived Human PDXDC1 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23042
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less