missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDE10A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531293
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531293 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531293 Supplier Invitrogen™ Supplier No. PA531293
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Recommended positive controls: HCT116, mouse brain, rat brain, Myc-DDK-tagged PDE10A-transfected 293T. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Various cellular responses are regulated by the second messengers cAMP and cGMP. Phosphodiesterases, such as PDE10A, eliminate cAMP- and cGMP-mediated intracellular signaling by hydrolyzing the cyclic nucleotide to the corresponding nucleoside 5-prime monophosphate.

Specifications

Antigen PDE10A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene PDE10A
Gene Accession No. Q8CA95, Q9QYJ6, Q9Y233
Gene Alias ADSD2; cAMP and cAMP-inhibited cGMP 3',5'-cyclic phosphodiesterase 10A; dJ416F21.1 (phosphodiesterase 10A); dual-substrate cAMP and cGMP, cyclic nucleotide phosphodiesterase; FLJ11894; FLJ25677; HSPDE10A; IOLOD; LINC00473; PDE10A; PDE10A19; PDE10A2 brain-specific isoform; Pde10a3; phosphodiesterase 10A; phosphodiesterase 10A1 (PDE10A1); phosphodiesterase 10A7 (PDE10A7); testis-specific phosphodiesterase PDE10A2
Gene Symbols PDE10A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human PDE10A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10846, 23984, 63885
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less