missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| PD 173074 | |
| 86°C | |
| MFCD08705327 | |
| 1-tert-butyl-3-2-4-diethylamino butyl amino-6-3,5-dimethoxyphenyl pyrido 2,3-d pyrimidin-7-yl urea, unii-a4tll8634y, 3-tert-butyl-1-2-4-diethylamino butyl amino-6-3,5-dimethoxyphenyl pyrido 2,3-d pyrimidin-7-yl urea, fgf/vegf receptor tyrosine kinase inhibitor, pd173074, 1-tert-butyl-3-2-4-diethylamino butylamino-6-3,5-dimethoxyphenyl pyrido 2,3-d pyrimidin-7-yl urea, 2fgi, 1-t-butyl-3-6-3,5-dimethoxyphenyl-2-4-diethylaminobutylamino-pyrido 2,3-d pyrimidin-7-yl urea, 1-tert-butyl-3-6-3,5-dimethoxy-phenyl-2-4-diethylamino-butylamino-pyrido 2,3-d pyrimidin-7-yl-urea | |
| CCN(CC)CCCCNC1=NC2=NC(=C(C=C2C=N1)C3=CC(=CC(=C3)OC)OC)NC(=O)NC(C)(C)C | |
| 523.682 | |
| CHEBI:63448 | |
| ≥95.0% (HPLC) |
| 219580-11-7 | |
| C28H41N7O3 | |
| 50 mg | |
| DXCUKNQANPLTEJ-UHFFFAOYSA-N | |
| 1-tert-butyl-3-[2-[4-(diethylamino)butylamino]-6-(3,5-dimethoxyphenyl)pyrido[2,3-d]pyrimidin-7-yl]urea | |
| 1401 | |
| 523.68 | |
| Crystalline Powder |
Chemical Identifiers
| 219580-11-7 | |
| 523.682 | |
| DXCUKNQANPLTEJ-UHFFFAOYSA-N | |
| 1401 | |
| 1-tert-butyl-3-[2-[4-(diethylamino)butylamino]-6-(3,5-dimethoxyphenyl)pyrido[2,3-d]pyrimidin-7-yl]urea |
| C28H41N7O3 | |
| MFCD08705327 | |
| 1-tert-butyl-3-2-4-diethylamino butyl amino-6-3,5-dimethoxyphenyl pyrido 2,3-d pyrimidin-7-yl urea, unii-a4tll8634y, 3-tert-butyl-1-2-4-diethylamino butyl amino-6-3,5-dimethoxyphenyl pyrido 2,3-d pyrimidin-7-yl urea, fgf/vegf receptor tyrosine kinase inhibitor, pd173074, 1-tert-butyl-3-2-4-diethylamino butylamino-6-3,5-dimethoxyphenyl pyrido 2,3-d pyrimidin-7-yl urea, 2fgi, 1-t-butyl-3-6-3,5-dimethoxyphenyl-2-4-diethylaminobutylamino-pyrido 2,3-d pyrimidin-7-yl urea, 1-tert-butyl-3-6-3,5-dimethoxy-phenyl-2-4-diethylamino-butylamino-pyrido 2,3-d pyrimidin-7-yl-urea | |
| CHEBI:63448 | |
| CCN(CC)CCCCNC1=NC2=NC(=C(C=C2C=N1)C3=CC(=CC(=C3)OC)OC)NC(=O)NC(C)(C)C |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator