accessibility menu, dialog, popup

UserName

CAS RN 219580-11-7

H3CNH3CNHNNNOCH3OCH3NHONHH3CCH3H3C

CAS RN 219580-11-7

IUPAC Name: 3-tert-butyl-1-(2-{[4-(diethylamino)butyl]amino}-6-(3,5-dimethoxyphenyl)pyrido[2,3-d]pyrimidin-7-yl)urea
Synonyms: 3-tert-butyl-1-(2-{[4-(diethylamino)butyl]amino}-6-(3,5-dimethoxyphenyl)pyrido[2,3-d]pyrimidin-7-yl)urea
Molecular Weight (g/mol): 523.68
Molecular Formula: C28H41N7O3
InChi Key: DXCUKNQANPLTEJ-UHFFFAOYSA-N
SMILES: CCN(CC)CCCCNC1=NC=C2C=C(C(NC(=O)NC(C)(C)C)=NC2=N1)C1=CC(OC)=CC(OC)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results