missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCSK5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150274
Description
PCSK5 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
PCSK5 belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. PCSK5 mediates posttranslational endoproteolytic processing for several integrin alpha subunits. It is thought to process prorenin, pro-membrane type-1 matrix metalloproteinase and HIV-1 glycoprotein gp160. Two alternatively spliced transcripts are described for this gene but only one has its full length nature known. The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein mediates posttranslational endoproteolytic processing for several integrin alpha subunits. It is thought to process prorenin, pro-membrane type-1 matrix metalloproteinase and HIV-1 glycoprotein gp160. Two alternatively spliced transcripts are described for this gene but only one has its full length nature known.Chemical Identifiers
| PCSK5 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q92824 | |
| 5125 | |
| Recombinant Proprotein Convertase Subtilisin/Kexin Type 5 (PCSK5) (Asp115-Met454). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Pcsk5 | |
| b2b1549Clo; b2b585Clo; PC5; PC5/6A; PC5A; PC6; PC6A; PCSK5; prohormone convertase 5; proprotein convertase 5; Proprotein convertase 6; proprotein convertase PC5; proprotein convertase subtilisin/kexin type 5; protease PC6; rPC5; SPC6; subtilase; Subtilisin/kexin-like protease PC5; subtilisin-like proprotein convertase 6 | |
| Pcsk5 | |
| Primary | |
| Protein A |
Specifications
| PCSK5 | |
| Polyclonal | |
| Unconjugated | |
| Pcsk5 | |
| b2b1549Clo; b2b585Clo; PC5; PC5/6A; PC5A; PC6; PC6A; PCSK5; prohormone convertase 5; proprotein convertase 5; Proprotein convertase 6; proprotein convertase PC5; proprotein convertase subtilisin/kexin type 5; protease PC6; rPC5; SPC6; subtilase; Subtilisin/kexin-like protease PC5; subtilisin-like proprotein convertase 6 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5125 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q92824 | |
| Pcsk5 | |
| Recombinant Proprotein Convertase Subtilisin/Kexin Type 5 (PCSK5) (Asp115-Met454). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |