missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PCR Product Pre-Sequencing Kit

Catalog No. 709951KT
Encompass
Change view
Click to view available options
No. of Reactions:
100
2000
500 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
709951KT 100
709961KT 500 Reactions
709971KT 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 709951KT Supplier Applied Biosystems™ Supplier No. 70995.1.KT

The PCR Product Pre-Sequencing Kit uses a novel, enzymatic clean-up method to pre-treat PCR products prior to sequencing, without any subsequent purification or separation steps.

  • Remove unincorporated primers and nucleotides in about 5 minutes
  • Prepare DNA sequencing samples with one pipetting step
  • Conserve precious PCR products with up to 100% recovery, regardless of amplicon length
  • Minimize spin column or bead clean-up

Specifications

Content And Storage Store at -5 to -30°C.
Description PCR Product Pre-Sequencing Kit, 100 rxns, Recombinant, PCR Purification, Uses a Novel, Enzymatic Clean-up Method to Pre-treat PCR Products Prior to Sequencing, without Any Subsequent Purification or Separation Steps
For Use With (Application) PCR Purification
No. of Reactions 100
Product Line ExoSAP-IT
Protein Form Recombinant
Quantity 100 reactions
Research Category Genetic Diseases
Shipping Condition Dry Ice