Learn More
Agilent BioTek PCR Microplate Holder
Stand for use with PCR microplates.
Supplier:  Agilent BioTek 6002076
Catalog No. BT6002076
Only null left
Explore available promotions
Description
Description
  • For PCR microplates such as MJ Research Part Number MLP-9601 and Nunc Part Number 259676
Microplates, washer dispensers, instruments, stands, accessories
Product Suggestions
Product Suggestions