Learn More
Invitrogen™ PCNA Monoclonal Antibody (PC10), Alexa Fluor™ Plus 555
Description
Description: The PC10 antibody reacts with human PCNA which is involved in the control of eukaryotic DNA replication and found in the nucleus. Applications Reported: This PC10 antibody has been reported for use in flow cytometric analysis, immunocytochemistry, western blotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections. Applications Tested: This PC10 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded tissue using high pH antigen retrieval and can be used at 20 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining.
Specifications
Specifications
| Antigen | PCNA |
| Applications | Immunohistochemistry (Paraffin), Multiplex Immunohistochemistry |
| Classification | Monoclonal |
| Clone | PC10 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor Plus 555 |
| Formulation | PBS with 0.5% BSA, 10% proprietary stabilizer and 0.05% sodium azide; pH 7.2 |
| Gene | Pcna |
| Gene Accession No. | P12004 |
| Gene Alias | ATLD2; cb16; Cyclin; DNA polymerase delta auxiliary protein; etID36690.10; fa28e03; fb36g03; HGCN8729; hypothetical protein LOC515499; MGC8367; Pcna; pcna protein; Pcna/cyclin; PCNAR; POL30; Proliferating cell nuclear antigen; wu:fa28e03; wu:fb36g03; YBR0811; YBR088C |
| Show More |