missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCDHGC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531382
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531382 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531382 Supplier Invitrogen™ Supplier No. PA531382
Only null left

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, Raji. Predicted reactivity: Mouse (99%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.

Specifications

Antigen PCDHGC4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene PCDHGC4
Gene Accession No. Q9Y5F7
Gene Alias PCDH-gamma-C4; PCDHGC4; protocadherin gamma subfamily C, 4; protocadherin gamma-C4
Gene Symbols PCDHGC4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Extracellular domain of human PCDHGC4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56098
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less