missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDHB15 Monoclonal Antibody (2E10)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524478
Description
Peptide Sequence: ELRLTLTAVD GGSPPRSGTV QILILVLDAN DNAPEFVQAL YEVQVPENSP VGSLVVKVSA RDLDTGTNGE ISYSLYYSSQ EIDKPFELSS LSGEIRLIKK.
Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintenance of specific neuronal connections in the brain.Chemical Identifiers
| PCDHB15 | |
| 1 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with no preservative; pH 7.4 | |
| Q9Y5E8 | |
| 56121 | |
| PCDHB15 (NP_061758, 207 a.a. ~ 306 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| Antibody |
| 2E10 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Monoclonal | |
| Liquid | |
| IgG2a κ | |
| Human | |
| PCDHB15 | |
| PCDHB15; PCDH-BETA15; PCDH-beta-15; protocadherin beta 15; protocadherin beta-15 | |
| PCDHB15 | |
| Primary | |
| Protein A |
Specifications
| PCDHB15 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9Y5E8 | |
| PCDHB15 | |
| PCDHB15 (NP_061758, 207 a.a. ~ 306 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2E10 | |
| Unconjugated | |
| PCDHB15 | |
| PCDHB15; PCDH-BETA15; PCDH-beta-15; protocadherin beta 15; protocadherin beta-15 | |
| Mouse | |
| Protein A | |
| RUO | |
| 56121 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |